siRNA Details
| Gene Symbol: Rpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053971.3 | NP_446423.2 | 1 | 124 | ||
| XM_006249377.1 | XP_006249439.1 | 2 | 143 | |||
| XM_006249378.2 | XP_006249440.1 | 2 | 177 | |||
| GenBank mRNA | BC061784.1 | - | 1 | 120 | ||
| BC078761.1 | - | 1 | 127 | 7123086 | ||
| BC097295.1 | - | 1 | 128 | 7446680 | ||
| FQ229484.1 | - | 2 | 142 | |||
| FQ229943.1 | - | 2 | 141 | |||
| FQ234880.1 | - | 2 | 141 | |||
| X87107.1 | - | 2 | 138 | |||

Rpl6
Genomic Map