siRNA Details
| Gene Symbol: RragB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053972.2 | NP_446424.1 | 9 | 1387 | ||
| XM_006256779.2 | XP_006256841.1 | 8 | 1401 | |||
| XM_008773066.1 | XP_008771288.1 | 8 | 1542 | |||
| XM_008773067.1 | XP_008771289.1 | 7 | 1458 | |||
| GenBank mRNA | BC078760.1 | - | 9 | 925 | 7123902 | |
| BC087698.1 | - | 9 | 1387 | 7314028 | ||
| Gene Symbol: LOC100909655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752022.3 | XP_003752070.1 | 1486 | |||
| XM_006256775.2 | XP_006256837.1 | 1401 | ||||
| XM_008773107.1 | XP_008771329.1 | 1542 | ||||
| XM_008773108.1 | XP_008771330.1 | 1458 | ||||
| XM_008773109.1 | XP_008771331.1 | 8 | 1542 | |||

RragB
Genomic Map