siRNA Details
| Gene Symbol: Rraga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053973.2 | NP_446425.1 | 1 | 307 | ||
| GenBank mRNA | BC061850.1 | - | 1 | 308 | 5621998 | |
| CK599679.1 | - | 1 | 307 | |||
| X85183.1 | - | 1 | 292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rraga| Gene Symbol: Rraga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053973.2 | NP_446425.1 | 1 | 307 | ||
| GenBank mRNA | BC061850.1 | - | 1 | 308 | 5621998 | |
| CK599679.1 | - | 1 | 307 | |||
| X85183.1 | - | 1 | 292 | |||