siRNA Details
| Gene Symbol: Cdh17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053977.2 | NP_446429.1 | 3 | 237 | ||
| XM_008763521.1 | XP_008761743.1 | 3 | 1691 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh17| Gene Symbol: Cdh17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053977.2 | NP_446429.1 | 3 | 237 | ||
| XM_008763521.1 | XP_008761743.1 | 3 | 1691 | |||