siRNA Details
| Gene Symbol: Arfrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053980.1 | NP_446432.1 | 5 | 436 | ||
| XM_006235718.2 | XP_006235780.1 | 5 | 432 | |||
| XM_006235720.2 | XP_006235782.1 | 5 | 389 | |||
| GenBank mRNA | X78603.1 | - | 5 | 436 | ||

Arfrp1
Genomic Map