siRNA Details
| Gene Symbol: Cd52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053983.1 | NP_446435.1 | 2 | 133 | ||
| GenBank mRNA | X76697.1 | - | 2 | 133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd52| Gene Symbol: Cd52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053983.1 | NP_446435.1 | 2 | 133 | ||
| GenBank mRNA | X76697.1 | - | 2 | 133 | ||