siRNA Details
| Gene Symbol: Gjb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053984.2 | NP_446436.1 | 1 | 1048 | ||
| XM_008763985.1 | XP_008762207.1 | 2 | 778 | |||
| XM_008763986.1 | XP_008762208.1 | 1 | 376 | |||
| GenBank mRNA | X76168.1 | - | 1 | 1048 | ||

Gjb4
Genomic Map