siRNA Details
| Gene Symbol: H3f3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053985.2 | NP_446437.1 | 4 | 1334 | ||
| GenBank mRNA | BC063159.1 | - | 4 | 1334 | 6921452 | |
| BC078759.1 | - | 4 | 1303 | 7112262 | ||
| FQ227664.1 | - | 1 | 658 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H3f3b| Gene Symbol: H3f3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053985.2 | NP_446437.1 | 4 | 1334 | ||
| GenBank mRNA | BC063159.1 | - | 4 | 1334 | 6921452 | |
| BC078759.1 | - | 4 | 1303 | 7112262 | ||
| FQ227664.1 | - | 1 | 658 | |||