siRNA Details
| Gene Symbol: Pou2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228368.2 | XP_006228430.2 | 10 | 975 | ||
| XM_006228371.2 | XP_006228433.2 | 10 | 993 | |||
| XM_006228372.2 | XP_006228434.2 | 10 | 975 | |||
| XM_008758885.1 | XP_008757107.1 | 10 | 1041 | |||
| GenBank mRNA | FQ225995.1 | - | 3 | 295 | ||
| L25863.1 | - | 10 | 878 | |||
| X67302.1 | - | 2 | 272 | |||

Pou2f2
Genomic Map