siRNA Details
Gene Symbol: Ubr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241570.2 | XP_006241632.2 | 262 | |||
XM_008765468.1 | XP_008763690.1 | 262 | ||||
XM_008765469.1 | XP_008763691.1 | 262 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ubr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241570.2 | XP_006241632.2 | 262 | |||
XM_008765468.1 | XP_008763690.1 | 262 | ||||
XM_008765469.1 | XP_008763691.1 | 262 |