siRNA Details
| Gene Symbol: Serpinb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130404.1 | NP_569088.1 | 6 | 735 | ||
| XM_006249629.1 | XP_006249691.1 | 7 | 815 | |||
| XM_008769420.1 | XP_008767642.1 | 7 | 791 | |||
| GenBank mRNA | AF105329.1 | - | 6 | 735 | ||
| FQ228843.1 | - | 7 | 796 | |||

Serpinb7
Genomic Map