siRNA Details
| Gene Symbol: Bdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053995.3 | NP_446447.2 | 6 | 726 | ||
| XM_006248478.2 | XP_006248540.1 | 6 | 782 | |||
| XM_008768691.1 | XP_008766913.1 | 5 | 564 | |||
| GenBank mRNA | BC085916.1 | - | 6 | 726 | 7108483 | |
| FQ211457.1 | - | 3 | 280 | |||
| FQ225409.1 | - | 1 | 53 | |||

Bdh1
Genomic Map