siRNA Details
| Gene Symbol: Kcnc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053997.4 | NP_446449.2 | 3, 4 | 2389 | ||
| XM_006228965.2 | XP_006229027.1 | 4, 5 | 2616 | |||
| XM_006228967.2 | XP_006229029.1 | 3, 4 | 2586 | |||
| GenBank mRNA | AY179603.1 | - | 3, 4 | 2389 | ||

Kcnc3
Genomic Map