siRNA Details
| Gene Symbol: Kcnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054000.2 | NP_446452.2 | 3 | 1428 | ||
| XM_008763474.1 | XP_008761696.1 | 2 | 586 | |||
| GenBank mRNA | M77482.1 | - | 3 | 1431 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001098528, BC146609.

Kcnb2
Genomic Map