siRNA Details
| Gene Symbol: B3gat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054003.1 | NP_446455.1 | 6 | 1170 | ||
| XM_006242733.2 | XP_006242795.1 | 6 | 1442 | |||
| XM_006242734.2 | XP_006242796.1 | 6 | 1431 | |||
| GenBank mRNA | D88035.1 | - | 6 | 1170 | ||

B3gat1
Genomic Map