siRNA Details
| Gene Symbol: Grifin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057187.2 | NP_476535.1 | 4 | 367 | ||
| XM_008768938.1 | XP_008767160.1 | 4 | 421 | |||
| GenBank mRNA | AF082160.1 | - | 4 | 367 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grifin| Gene Symbol: Grifin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057187.2 | NP_476535.1 | 4 | 367 | ||
| XM_008768938.1 | XP_008767160.1 | 4 | 421 | |||
| GenBank mRNA | AF082160.1 | - | 4 | 367 | ||