siRNA Details
| Gene Symbol: Hivep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105751.1 | NP_001099221.1 | 10 | 7184 | ||
| GenBank mRNA | X54250.1 | - | 5 | 996 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hivep1| Gene Symbol: Hivep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105751.1 | NP_001099221.1 | 10 | 7184 | ||
| GenBank mRNA | X54250.1 | - | 5 | 996 | ||