siRNA Details
| Gene Symbol: Cuzd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054005.1 | NP_446457.1 | 8 | 1850 | ||
| GenBank mRNA | AF022147.1 | - | 8 | 1850 | ||
| AF167170.1 | - | 8 | 1616 | |||
| BC090345.1 | - | 8 | 1596 | |||
| BC107927.1 | - | 1651 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cuzd1| Gene Symbol: Cuzd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054005.1 | NP_446457.1 | 8 | 1850 | ||
| GenBank mRNA | AF022147.1 | - | 8 | 1850 | ||
| AF167170.1 | - | 8 | 1616 | |||
| BC090345.1 | - | 8 | 1596 | |||
| BC107927.1 | - | 1651 | ||||