siRNA Details
| Gene Symbol: Sh3bp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054011.1 | NP_446463.2 | 3 | 258 | ||
| GenBank mRNA | AB027562.1 | - | 3 | 258 | ||
| FQ210473.1 | - | 3 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3bp5| Gene Symbol: Sh3bp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054011.1 | NP_446463.2 | 3 | 258 | ||
| GenBank mRNA | AB027562.1 | - | 3 | 258 | ||
| FQ210473.1 | - | 3 | 385 | |||