siRNA Details
| Gene Symbol: Tenm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020088.1 | NP_064473.1 | 24 | 5324 | ||
| GenBank mRNA | AF086607.1 | - | 24 | 5324 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tenm2| Gene Symbol: Tenm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020088.1 | NP_064473.1 | 24 | 5324 | ||
| GenBank mRNA | AF086607.1 | - | 24 | 5324 | ||