siRNA Details
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130739.1 | NP_570095.1 | 18 | 1806 | ||
GenBank mRNA | S56508.1 | - | 18 | 1806 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130739.1 | NP_570095.1 | 18 | 1806 | ||
GenBank mRNA | S56508.1 | - | 18 | 1806 |