siRNA Details
| Gene Symbol: Acsl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130739.1 | NP_570095.1 | 6 | 697 | ||
| XM_006246221.2 | XP_006246283.1 | 6 | 858 | |||
| XM_006246222.2 | XP_006246284.1 | 6 | 858 | |||
| XM_006246223.2 | XP_006246285.1 | 6 | 1015 | |||
| XM_006246224.2 | XP_006246286.1 | 6 | 738 | |||
| GenBank mRNA | AY625254.1 | - | 5 | 558 | ||
| D10041.1 | - | 6 | 678 | |||
| S56508.1 | - | 6 | 697 | |||

Acsl6
Genomic Map