siRNA Details
| Gene Symbol: Dnah9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727722.4 | XP_002727768.2 | 37 | 7383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnah9| Gene Symbol: Dnah9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727722.4 | XP_002727768.2 | 37 | 7383 | ||