siRNA Details
| Gene Symbol: Ppp2r2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057116.1 | NP_476457.1 | 565 | |||
| GenBank mRNA | D38261.1 | - | 565 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp2r2c| Gene Symbol: Ppp2r2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057116.1 | NP_476457.1 | 565 | |||
| GenBank mRNA | D38261.1 | - | 565 | |||