siRNA Details
| Gene Symbol: Cntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057118.2 | NP_476459.1 | 17 | 2161 | ||
| XM_006242177.2 | XP_006242239.1 | 17 | 2176 | |||
| GenBank mRNA | D38492.1 | - | 17 | 2165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntn1| Gene Symbol: Cntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057118.2 | NP_476459.1 | 17 | 2161 | ||
| XM_006242177.2 | XP_006242239.1 | 17 | 2176 | |||
| GenBank mRNA | D38492.1 | - | 17 | 2165 | ||