siRNA Details
| Gene Symbol: Nudt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057120.1 | NP_476461.1 | 3 | 601 | ||
| XM_006248911.2 | XP_006248973.1 | 4 | 652 | |||
| GenBank mRNA | D49977.1 | - | 3 | 601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt1| Gene Symbol: Nudt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057120.1 | NP_476461.1 | 3 | 601 | ||
| XM_006248911.2 | XP_006248973.1 | 4 | 652 | |||
| GenBank mRNA | D49977.1 | - | 3 | 601 | ||