siRNA Details
| Gene Symbol: Nr0b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057133.1 | NP_476474.1 | 2 | 767 | ||
| GenBank mRNA | BC088117.1 | - | 2 | 754 | 7324177 | |
| D86580.1 | - | 2 | 767 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr0b2| Gene Symbol: Nr0b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057133.1 | NP_476474.1 | 2 | 767 | ||
| GenBank mRNA | BC088117.1 | - | 2 | 754 | 7324177 | |
| D86580.1 | - | 2 | 767 | |||