siRNA Details
| Gene Symbol: Chrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057134.1 | NP_476475.1 | 8 | 954 | ||
| XM_006248556.2 | XP_006248618.1 | 8 | 1336 | |||
| XM_006248557.2 | XP_006248619.1 | 8 | 1360 | |||
| GenBank mRNA | FQ222446.1 | - | 8 | 993 | ||
| FQ223218.1 | - | 8 | 993 | |||

Chrd
Genomic Map