siRNA Details
| Gene Symbol: Ebp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057137.1 | NP_476478.1 | 5 | 755 | ||
| XM_006256699.2 | XP_006256761.1 | 5 | 780 | |||
| GenBank mRNA | AF071501.1 | - | 5 | 755 | ||
| FQ209542.1 | - | 3 | 337 | |||
| FQ219951.1 | - | 3 | 337 | |||
| FQ230059.1 | - | 3 | 337 | |||

Ebp
Genomic Map