siRNA Details
| Gene Symbol: Hnrnpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057139.2 | NP_476480.2 | 12 | 2272 | ||
| XM_006250311.2 | XP_006250373.1 | 12 | 2648 | |||
| GenBank mRNA | BC072529.1 | - | 12 | 2272 | 7096906 | |
| D14048.1 | - | 12 | 2751 | |||

Hnrnpu
Genomic Map