siRNA Details
| Gene Symbol: Hnrnpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057141.1 | NP_476482.1 | 14 | 1261 | ||
| XM_006253552.2 | XP_006253614.1 | 16 | 1668 | |||
| XM_006253553.2 | XP_006253615.1 | 15 | 1364 | |||
| XM_006253554.2 | XP_006253616.1 | 16 | 1668 | |||
| XM_006253555.2 | XP_006253617.1 | 15 | 1596 | |||
| GenBank mRNA | BC061867.1 | - | 15 | 1348 | 5598872 | |
| D17711.1 | - | 14 | 1261 | |||

Hnrnpk
Genomic Map