siRNA Details
| Gene Symbol: Lrrc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057142.1 | NP_476483.1 | 4, 5 | 499 | ||
| GenBank mRNA | EU348640.1 | - | 3, 4 | 422 | ||
| EU348641.1 | - | 3, 4 | 423 | |||
| EU348642.1 | - | 4, 5 | 499 | |||
| EU348643.1 | - | 4, 5 | 499 | |||
| EU348644.1 | - | 4, 5 | 499 | |||
| U66707.1 | - | 4, 5 | 499 | |||

Lrrc7
Genomic Map