siRNA Details
| Gene Symbol: Sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057147.2 | NP_476488.2 | 6 | 712 | ||
| XM_006248419.2 | XP_006248481.1 | 7 | 914 | |||
| XM_006248420.2 | XP_006248482.1 | 6 | 949 | |||
| XM_008768692.1 | XP_008766914.1 | 6 | 1161 | |||
| XM_008768693.1 | XP_008766915.1 | 6 | 1163 | |||
| GenBank mRNA | BC081746.1 | - | 6 | 712 | 7135589 | |

Sec22a
Genomic Map