siRNA Details
| Gene Symbol: Txnip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008767.1 | NP_001008767.1 | 3 | 529 | ||
| GenBank mRNA | BC088411.1 | - | 3 | 529 | 7192327 | |
| U30789.1 | - | 3 | 1314 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txnip| Gene Symbol: Txnip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008767.1 | NP_001008767.1 | 3 | 529 | ||
| GenBank mRNA | BC088411.1 | - | 3 | 529 | 7192327 | |
| U30789.1 | - | 3 | 1314 | |||