siRNA Details
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100474.1 | NP_001093944.1 | 15, 16 | 1739 | ||
GenBank mRNA | AF410816.1 | - | 15, 16 | 1739 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100474.1 | NP_001093944.1 | 15, 16 | 1739 | ||
GenBank mRNA | AF410816.1 | - | 15, 16 | 1739 |