siRNA Details
| Gene Symbol: Ccnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100474.1 | NP_001093944.1 | 16 | 1805 | ||
| GenBank mRNA | AF410816.1 | - | 16 | 1805 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccnf| Gene Symbol: Ccnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100474.1 | NP_001093944.1 | 16 | 1805 | ||
| GenBank mRNA | AF410816.1 | - | 16 | 1805 | ||