siRNA Details
| Gene Symbol: Tbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004198.1 | NP_001004198.1 | 7 | 1106 | ||
| XM_006227980.2 | XP_006228042.1 | 7 | 1049 | |||
| XM_008758747.1 | XP_008756969.1 | 7 | 1049 | |||
| XM_008758748.1 | XP_008756970.1 | 7 | 1257 | |||
| GenBank mRNA | BC081939.1 | - | 7 | 1106 | 7115389 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013684, BC050136, BC016476, D01034, DQ906043, and others.

Tbp
Genomic Map