siRNA Details
Gene Symbol: Klhl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057191.1 | NP_476539.1 | 1 | 1063 | ||
GenBank mRNA | AJ293948.1 | - | 1 | 1063 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057191.1 | NP_476539.1 | 1 | 1063 | ||
GenBank mRNA | AJ293948.1 | - | 1 | 1063 |