siRNA Details
| Gene Symbol: Klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057191.1 | NP_476539.1 | 1 | 660 | ||
| GenBank mRNA | AJ293948.1 | - | 1 | 660 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl41| Gene Symbol: Klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057191.1 | NP_476539.1 | 1 | 660 | ||
| GenBank mRNA | AJ293948.1 | - | 1 | 660 | ||