siRNA Details
| Gene Symbol: Il10ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057193.2 | NP_476541.2 | 7 | 1616 | ||
| XM_006242864.2 | XP_006242926.1 | 7 | 1794 | |||
| GenBank mRNA | AJ305049.1 | - | 7 | 1616 | ||
| FQ223464.1 | - | 7 | 1637 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il10ra| Gene Symbol: Il10ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057193.2 | NP_476541.2 | 7 | 1616 | ||
| XM_006242864.2 | XP_006242926.1 | 7 | 1794 | |||
| GenBank mRNA | AJ305049.1 | - | 7 | 1616 | ||
| FQ223464.1 | - | 7 | 1637 | |||