siRNA Details
| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057202.1 | NP_476550.1 | 4, 5 | 386 | ||
| XM_006254005.1 | XP_006254067.1 | 4, 5 | 817 | |||
| GenBank mRNA | DQ309275.1 | - | 4, 5 | 386 | ||
| FQ226753.1 | - | 4, 5 | 780 | |||
| FQ227346.1 | - | 2, 3 | 177 | |||
| FQ230391.1 | - | 4, 5 | 809 | |||
| FQ232221.1 | - | 2, 3 | 177 | |||
| FQ233216.1 | - | 4, 5 | 789 | |||
| FQ234099.1 | - | 2, 3 | 177 | |||
| FQ234494.1 | - | 2, 3 | 177 | |||
| FQ235192.1 | - | 2, 3 | 177 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008448, BC025864, BC040800, AK185672, L27153, and others.

Kif5b
Genomic Map