siRNA Details
| Gene Symbol: Ptpn23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057204.2 | NP_476552.2 | 13 | 1087 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn23| Gene Symbol: Ptpn23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057204.2 | NP_476552.2 | 13 | 1087 | ||