siRNA Details
| Gene Symbol: Uba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057205.2 | NP_476553.1 | 7, 8 | 468 | ||
| XM_006236936.2 | XP_006236998.1 | 6, 7 | 451 | |||
| GenBank mRNA | AF336829.1 | - | 7, 8 | 477 | ||
| BC081743.1 | - | 7, 8 | 468 | 7124631 | ||
| FQ234407.1 | - | 6, 7 | 427 | |||

Uba3
Genomic Map