siRNA Details
| Gene Symbol: Pnliprp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057206.1 | NP_476554.1 | 9 | 1083 | ||
| GenBank mRNA | L09216.1 | - | 9 | 1083 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnliprp2| Gene Symbol: Pnliprp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057206.1 | NP_476554.1 | 9 | 1083 | ||
| GenBank mRNA | L09216.1 | - | 9 | 1083 | ||