siRNA Details
| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057207.3 | NP_476555.1 | 9 | 1802 | ||
| XM_008759492.1 | XP_008757714.1 | 9 | 2521 | |||
| XM_008759493.1 | XP_008757715.1 | 9 | 2899 | |||
| XM_008759494.1 | XP_008757716.1 | 9 | 3421 | |||
| XM_008759495.1 | XP_008757717.1 | 9 | 2517 | |||
| GenBank mRNA | L10362.1 | - | 9 | 1802 | ||

Sv2b
Genomic Map