siRNA Details
| Gene Symbol: Rps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005528.1 | NP_001005528.1 | 3, 4 | 314 | ||
| GenBank mRNA | BC103658.1 | - | 3, 4 | 319 | 7446085 | |
| FQ210084.1 | - | 3, 4 | 332 | |||
| FQ217008.1 | - | 3, 4 | 320 | |||
| FQ217453.1 | - | 3, 4 | 304 | |||
| FQ221194.1 | - | 3, 4 | 335 | |||
| FQ221611.1 | - | 3, 4 | 334 | |||
| FQ221873.1 | - | 3, 4 | 333 | |||
| FQ228420.1 | - | 3, 4 | 335 | |||

Rps25
Genomic Map