siRNA Details
| Gene Symbol: Rps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005528.1 | NP_001005528.1 | 4 | 362 | ||
| GenBank mRNA | BC103658.1 | - | 4 | 367 | 7446085 | |
| FQ210084.1 | - | 4 | 380 | |||
| FQ217008.1 | - | 4 | 368 | |||
| FQ217453.1 | - | 4 | 352 | |||
| FQ221194.1 | - | 4 | 383 | |||
| FQ221611.1 | - | 4 | 382 | |||
| FQ221873.1 | - | 4 | 381 | |||
| FQ228420.1 | - | 4 | 383 | |||

Rps25
Genomic Map