siRNA Details
| Gene Symbol: Rps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078617.3 | NP_511172.1 | 4 | 403 | ||
| GenBank mRNA | BC058134.1 | - | 4 | 407 | 6918484 | |
| FQ221811.1 | - | 4 | 395 | |||
| FQ222488.1 | - | 4 | 393 | |||
| FQ222494.1 | - | 4 | 397 | |||
| FQ222496.1 | - | 4 | 394 | |||
| FQ222641.1 | - | 4 | 395 | |||
| FQ223677.1 | - | 4 | 395 | |||
| FQ223812.1 | - | 4 | 395 | |||
| FQ228659.1 | - | 4 | 395 | |||
| FQ228931.1 | - | 4 | 395 | |||

Rps23
Genomic Map