siRNA Details
Gene Symbol: Lama5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191609.1 | NP_001178538.1 | 67 | 9157 | ||
GenBank mRNA | FQ223406.1 | - | 13 | 1682 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lama5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191609.1 | NP_001178538.1 | 67 | 9157 | ||
GenBank mRNA | FQ223406.1 | - | 13 | 1682 |