siRNA Details
| Gene Symbol: Slc8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078619.1 | NP_511174.1 | 6, 7 | 2017 | ||
| GenBank mRNA | U08141.1 | - | 6, 7 | 2017 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc8a2| Gene Symbol: Slc8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078619.1 | NP_511174.1 | 6, 7 | 2017 | ||
| GenBank mRNA | U08141.1 | - | 6, 7 | 2017 | ||